Ethylacetat ≥99,5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

23882.296EA 28.5 EUR
23882.296 23882.321 23882.467 VWRC23882.550 23882.330 23882.310 23882.360 23882.950
Ethylacetat ≥99,5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens
Ethylacetat
Formel: CH₃COOC₂H₅
Siedepunkt: 77,1 °C (1013 hPa)
Schmelzpunkt: –83 °C
Dichte: 0,902 g/cm³ (20 °C)
Flammpunkt: –4 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009171
CAS-Nummer: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (on anhydrous substance) Min. 99.5 %
IR Spectrum Passes test
Organic impurities Passes test
Acidity Max. 0.0006 meq/g
Boiling point 76.0 - 77.5 °C
Colouration Max. 10 APHA
Density (20/4) 0.899 - 0.901
Density (20/20) 0.901 - 0.904
Substances coloured by H2SO4 Max. 60 APHA
Ethanol Max. 0.1 %
Evaporation residue Max. 10 ppm
Methanol Max. 0.1 %
Methyl acetate Max. 0.1 %
iso-Propyl acetate Max. 0.06 %
n-Propyl acetate Max. 0.5 %
Water Max. 0.03 %
Al (Aluminium) Max. 0.1 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.05 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR