Curcumin (aus Curcuma longa) ≥80% (durch HPLC), Calbiochem®, Sigma-Aldrich®

Lieferant: Merck Millipore Calbiochem

Synonyme: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione

M-Clarity™ Qualitätsstufe = MQ100

239802-100
239802-100EA 71.5 EUR
239802-100
Curcumin (aus Curcuma longa) ≥80% (durch HPLC), Calbiochem®, Sigma-Aldrich®
Curcumin

A cell-permeable and irreversible antitumor and anti-inflammatory agent that acts as an inhibitor of 5-lipoxygenase (IC₅₀ = 8 µM) and cyclooxygenase (IC₅₀ = 52 µM).

Formel: C₂₁H₂₀O₆
Molecular Weight: 368,39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008365
CAS-Nummer: 458-37-7
EINECS: 207-280-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR