Cyclopamine V. californicum 97 HPLC_ASSAY_METHOD, Millipore®
Lieferant: Merck Millipore Calbiochem
|
Achtung
|
Synonyme:
(2'R,3S,3'R,3a'S,6aS,6bS,6'S,7a'R,11aS,11bR)-3',6',10,11b-tetramethyl-1,2,3,3a',4,4',5',6,6a,6b,6',7,7',7a',8,11,11a,11b-octadecahydro-3'H-spiro[benzo[a]fluorene-9,2'-furo[3,2-b]pyridin]-3-ol
239803-1
239803-5MG
239803-1EA
280
EUR
239803-1
239803-5MG
Cyclopamine V. californicum 97 HPLC_ASSAY_METHOD, Millipore®
Cyclopamine
Cyclopamine, V. californicum, is a cell-permeable steroidal alkaloid & cholesterol mimic that specifically antagonises sonic hedgehog signaling through direct interaction with Smo.
Verpackung: Packaged under inert gas.
Formel:
C₂₇H₄₁NO₂ Molecular Weight: 411.63 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD09878269 CAS-Nummer: 4449-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...