Test tube holder, stainless steel
Lieferant: HAMMACHER KARL
HWN322-13
241-0233EA 78.6 EUR
241-0233
Test tube holder, stainless steel
Zangen
Test tube holder made from WIRONIT steel (18/12 stainless steel with 18% chrome and 10 to 12% nickel).