Doppelmuffen
Lieferant: Lenz Laborglas GmbH & CO.KG
06250000
241-2550EA 57.8 EUR
241-2550
Doppelmuffen
Klemmen Stativmuffen
Bosshead made from tempered cast iron. For attaching reaction vessel clamps to support stands.
  • Clasping range up to 25 mm
  • Fast and easy assembly and disassembly via large brass tommy screws
  • Robust, durable
Standards: DIN 12895