Retort clamp, four pronged
Lieferant: BOCHEM
5423
241-7423EA 171 EUR
241-7423
Retort clamp, four pronged
Klemmen Klemmen für den allgemeinen Gebrauch
This four pronged retort clamp is made of stainless steel.
  • Fingers are coated with silicone to protect clamped items