Chromium(III) phosphate tetrahydrate

Lieferant: Thermo Fisher Scientific
Achtung

24110.09 24110.18
24110.09EA 60.1 EUR
24110.09 24110.18
Chromium(III) phosphate tetrahydrate
Chromium(III) phosphate tetrahydrate

CAS No.: 10101-59-4 Molecular Formula: CrPO4.4H2O Formula Weight: 219.04 (146.97anhy) Physical Form: Powder Density: 2.26 MDL No.: MFCD00016049


Applications: Catalyst, pigment

Formel: CrH₈O₈P
Dichte: 2,26
Lagertemperatur: Raumtemperatur
MDL: MFCD00016049
CAS-Nummer: 10101-59-4
EINECS: 232-141-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR