Eisen(III)-nitrat Nonahydrat 99,0-101,0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens

Lieferant: VWR Chemicals
Achtung

Synonyme: Eisentrinitrat Nonahydrat

24175.233EA 51.1 EUR
24175.233 24175.290 24175.260
Eisen(III)-nitrat Nonahydrat 99,0-101,0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
Eisen (III) nitrat Nonahydrat
Formel: Fe(NO₃)₃·9H₂O
Molecular Weight: 404 g/mol
Siedepunkt: 125
Schmelzpunkt: 47
Dichte: 1.68 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149708
CAS-Nummer: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 5.1 III
Merck Index: 13,04057

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 101.0 %
Free acid Max. 0.05 meq/g
Insolubility in water Max. 0.02 %
Not precipitated by NH4OH (as SO4) Max. 0.1 %
Cl (Chloride) Max. 5 ppm
PO4 (Phosphate) Max. 50 ppm
SO4 (Sulphate) Max. 50 ppm
Ca (Calcium) Max. 50 ppm
Cu (Copper) Max. 50 ppm
K (Potassium) Max. 50 ppm
Mg (Magnesium) Max. 10 ppm
Mn (Manganese) Max. 0.02 %
Na (Sodium) Max. 50 ppm
Pb (Lead) Max. 10 ppm
Zn (Zinc) Max. 10 ppm
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR