Aktivkohle, Pulver, DARCO® -100 mesh, Fluka™

Lieferant: Honeywell Chemicals
Achtung

Synonyme: Carbon activated, Activated carbon, Activated charcoal

242276-1KG
242276-1KGEA 313.32 EUR
242276-1KG
Aktivkohle, Pulver, DARCO® -100 mesh, Fluka™
Aktivkohle

  • DARCO is a registered trademark of Norit Americas, Inc.

Formel: C
Molecular Weight: 12.01 g/mol
Schmelzpunkt: 3550
Lagertemperatur: Raumtemperatur
MDL: MFCD00133992
CAS-Nummer: 7440-44-0
EINECS: 231-153-3
REACH: 01-2119488894-16
Merck Index: 13,01818

Bestellen

Spezifikation Testergebnisse

Particle Size −100 mesh
Appearance complying
Loss on drying (105°C) Max. 12 %
Methylene blue adsorption Min. 15 g/100g
Water soluble matter Max. 0.50 %
In acid soluble iron (Fe) Max. 200 ppm
Vapor Pressure <0.1 mmHg ( 20 °C)
Resistivity 1375 μΩ-cm, 20°C (graphite)
Form powder
Autoignition Temperature 842 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR