Magnesiumacetat Tetrahydrat, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
Synonyme:
Essigsäure Magnesiumsalz-Tetrahydrat, Magnesiumdi(acetat) Tetrahydrat, Essigsäure Magnesiumsalz Tetrahydrat, Magnesiumdi(acetat)-Tetrahydrat
2424-01EA
376.7
EUR
2424-01
Magnesiumacetat Tetrahydrat, BAKER ANALYZED® ACS, J.T.Baker®
Magnesiumacetat Tetrahydrat
Formel:
(H₃CCOO)₂Mg·4H₂O Schmelzpunkt: 80 °C Dichte: 1,454 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149214 CAS-Nummer: 16674-78-5 EINECS: 205-554-9 Merck Index: 13,05676 |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | 98.0 - 102.0 % |
Assay ((CH₃COO)₂Mg·4H₂O) | 98.0 - 102.0 % |
Insoluble Matter | ≤0.005 % |
pH of 5% Solution at 25°C | 6.5 - 8.5 |
Chloride (Cl) | ≤0.001 % |
Sulfate (SO₄) | ≤0.005 % |
Barium (Ba) | ≤0.001 % |
Calcium (Ca) | ≤0.01 % |
Manganese (Mn) | ≤0.001 % |
Potassium (K) | ≤0.005 % |
Sodium (Na) | ≤0.005 % |
Strontium (Sr) | ≤0.005 % |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤5 ppm |
Appearance (colorless, deliquescent solid) | |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...