Eisen(II)sulfat Hydrat ≥84%, GPR RECTAPUR®
Lieferant: VWR Chemicals
|
Achtung
|
Synonyme:
Ferrous sulphate hydrate
24242.292EA
68.8
EUR
24242.292
Eisen(II)sulfat Hydrat ≥84%, GPR RECTAPUR®
Eisen(II)sulfat Hydrat
Formel:
FeSO₄·H₂O Molecular Weight: 169,93 g/mol Schmelzpunkt: ∼300 °C Dichte: 2,97 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149722 CAS-Nummer: 13463-43-9 EINECS: 231-753-5 |
Spezifikation Testergebnisse
Assay (on anhydrous substance) | Min. 84 % |
Heavy metals (as Pb) | Max. 50 ppm |
Insolubility in water | Max. 0.05 % |
As (Arsenic) | Max. 2 ppm |
Fe (III) (Iron) | Max. 0.4 % |
Pb (Lead) | Max. 10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...