Magnesiumacetat Tetrahydrat 98,0-102,0%, Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
Synonyme:
Essigsäure Magnesiumsalz-Tetrahydrat, Magnesiumdi(acetat) Tetrahydrat, Essigsäure Magnesiumsalz Tetrahydrat, Magnesiumdi(acetat)-Tetrahydrat
2425-07EA
0
EUR
2425-07
Magnesiumacetat Tetrahydrat 98,0-102,0%, Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Magnesiumacetat Tetrahydrat
Formel:
(H₃CCOO)₂Mg·4H₂O Molecular Weight: 214,46 g/mol Dichte: 1,45 g/cm³ |
MDL:
MFCD00149214 CAS-Nummer: 16674-78-5 |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | — |
For Laboratory,Research,or Manufacturing Use | — |
ACS - Assay ((CH₃COO)₂Mg·4H₂O) | 98.0 - 102.0 % |
ACS - Insoluble Matter | ≤0.005 % |
ACS - pH of 5% Solution at 25°C | 6.5 - 8.5 |
ACS - Chloride (Cl) | ≤0.001 % |
ACS - Sulfate (SO₄) | ≤0.005 % |
ACS - Barium (Ba) | ≤0.001 % |
ACS - Calcium (Ca) | ≤0.01 % |
ACS - Manganese (Mn) | ≤0.001 % |
ACS - Potassium (K) | ≤0.005 % |
ACS - Sodium (Na) | ≤0.005 % |
ACS - Strontium (Sr) | ≤0.005 % |
ACS - Endotoxin Concentration (2.5 EU/g max.) | Passes Test |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...