Eisen(III)sulfat Hydrat, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

24252.296EA 174 EUR
24252.296
Eisen(III)sulfat Hydrat, AnalaR NORMAPUR® analytisches Reagens
Eisen(III)sulfat Hydrat
Formel: Fe₂(SO₄)₃·xH₂O
Dichte: 3,1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149714
CAS-Nummer: 15244-10-7
EINECS: 233-072-9
Merck Index: 13,04065

Bestellen

Spezifikation Testergebnisse

Assay (calculated as Fe(III)) 21.0 - 23.0 %
Insolubility in diluted HCl Max. 100 ppm
Cl (Chloride) Max. 200 ppm
NO3 (Nitrate) Max. 0.5 %
PO4 (Phosphate) Max. 30 ppm
As (Arsenic) Max. 5 ppm
Cu (Copper) Max. 20 ppm
Fe (II) (Iron) Max. 0.02 %
Zn (Zinc) Max. 50 ppm
Ba (Barium) Max. 2 ppm
Mn (Manganese) Max. 0.1 %
Pb (Lead) Max. 6 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR