D(+)-Glucose Monohydra Ph. Eur.

Lieferant: VWR Chemicals

Synonyme: Dextrose Monohydrat, Traubenzucker Monohydrat

24369.290EA 66.8 EUR
24369.290 24369.461
D(+)-Glucose Monohydra Ph. Eur.
D(+)-Glucose Monohydrat

Je nach Bedingungen kann dieses Produkt apyrogen sein

Formel: C₆H₁₂O₆·H₂O
Schmelzpunkt: 83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149450
CAS-Nummer: 14431-43-7
EINECS: 200-075-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9ea2af2ca62ca94d86809070593a60aafd8cd1b&vid=e549b499dca7fb95811640efad1429268c8f6d8b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Residual solvents Passes test
Appearance Weisses kristallin.Pulver
Identification B Passes test
Identification C Passes test
Solution S Passes test
Appearance of solution Passes test
Acidity or alkalinity Passes test
Spec. opt. rotation (calc. on anhydrous) 52.5 - 53.3 deg
Foreign sugars, soluble starch, dextrins Passes test
SO3 (as SO2) Max. 15 ppm
Cl (Chloride) Max. 125 ppm
SO4 (Sulphate) Max. 200 ppm
As (Arsenic) Max. 1 ppm
Ba (Barium) Passes test
Ca (Calcium) Max. 200 ppm
Lead in sugars Max. 0.5 ppm
Water 7 - 9.5 %
Sulphated ash Max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR