Glycerin 87% 86%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals

Synonyme: 1,2,3-Propantriol, Glycerol

24385.295EA 106 EUR
24385.295 24385.320 24385.460
Glycerin 87% 86%, AnalaR NORMAPUR® analytisches Reagens
Glycerin
Formel: HOCH₂CH(OH)CH₂OH
Lagertemperatur: Raumtemperatur
MDL: MFCD00004722
CAS-Nummer: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Gehalt 83.5 - 88.0%
Acidity Max. 0.0004 meq/g
Alkalinity Max. 0.0003 meq/g
Density (20/4) 1.220 - 1.230
Esters of fatty acids (as C15H26O6) Max. 0.05 %
Glyceraldehyde Max. 30 ppm
Heavy metals (as Pb) Max. 1 ppm
Ignition residue (SO4) Max. 50 ppm
Cl (Chloride) Max. 1 ppm
NH4 (Ammonium) Max. 5 ppm
SO4 (Sulphate) Max. 5 ppm
Fe (Iron) Max. 1 ppm
Water 13 - 16 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR