Kieselgel, Davisil® Grade 12 pore size 150 Å, 60-200 mesh, high_purity, Supelco®

Lieferant: Merck

243981-2.5KG 243981-500G
243981-2.5KGEA 470 EUR
243981-2.5KG 243981-500G
Kieselgel, Davisil® Grade 12 pore size 150 Å, 60-200 mesh, high_purity, Supelco®
Silica gel
Formel: SiO₂
Molecular Weight: 60.08 g/mol
Siedepunkt: 2230 °C
Schmelzpunkt: > 1600 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 63231-67-4
EINECS: 231-545-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=861a98e5d9c23ce015dc5d3927c4e44df6690433&vid=e8d3ff63d9b9e9a21c4459655ec4da368565da34&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR