n-Heptan ≥99,0%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

24551.290EA 34.4 EUR
24551.290 24551.324 24551.368 24551.461 VWRC24551.550
n-Heptan ≥99,0%, AnalaR NORMAPUR® analytisches Reagens
n-Heptan
Formel: H₃C(CH₂)₅CH₃
Siedepunkt: 98,4 °C (1013 hPa)
Schmelzpunkt: –90,6 °C
Dichte: 0,6838 g/cm³ (20 °C)
Flammpunkt: –4 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009544
CAS-Nummer: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
REACH: 01-2119457603-38
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay Min. 99.0 %
IR Spectrum Passes test
Acidity Max. 0.0001 meq/g
Alkalinity Max. 0.0001 meq/g
Boiling point 98 - 99 °C
Bromine value Max. 0.5
Colouration Max. 10 APHA
Density (20/4) 0.681 - 0.685
Substances coloured by H2SO4 Max. 90 APHA
Aromatics (V/V) Max. 0.1 %
Evaporation residue Max. 10 ppm
Total S (as SO4) Max. 10 ppm
Water Max. 100 ppm
Al (Aluminium) Max. 0.1 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.1 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 0.1 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.02 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR