Silikonöl, technical Oilbath, VWR®

Lieferant: VWR Chemicals

Synonyme: Polyphenyl-methylsiloxane

24610.363EA 233 EUR
24610.363
Silikonöl, technical Oilbath, VWR®
Silikonöl

Verwendbar zwischen 100 und 200°C.


  • Essentially non-toxic
  • Environmentally safe
  • Non-halogenated
  • High thermal stability and oxidation resistance



Typical values (without guarantee)
• Appearance: crystal clear liquid
• Density (25°C): 0.96 kg/dm3
• Water content: about 30 ppm
• Viscosity (25 °C): 50 mm2/s
• Flash point (closed cup): 260 °C
• Fire point (open cup): 370 °C

Lagertemperatur: Raumtemperatur
MDL: MFCD00132673
CAS-Nummer: 68083-14-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR