Poly(chlortrifluorethylen) 500, VOLTALEF®
Lieferant: VWR Chemicals
24625.182EA
0
EUR
24625.182
Poly(chlortrifluorethylen) 500, VOLTALEF®
Poly(chlortrifluorethylen) 500
Formel:
(C₂ClF₃)n Molecular Weight: 500 g/mol |
CAS-Nummer:
9002-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...