Hydroxylammoniumchlorid ≥99,0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens
Lieferant: VWR Chemicals
|
Achtung
|
24708.235EA
105
EUR
24708.235
24708.292
Hydroxylammoniumchlorid ≥99,0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens
Hydroxylammoniumchlorid
Formel:
NH₂OH·HCl Siedepunkt: 306 °C (1013 hPa) Schmelzpunkt: 151 °C Dichte: 1,67 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051089 CAS-Nummer: 5470-11-1 EINECS: 226-798-2 UN: 2923 ADR: 8 (6.1),III Merck Index: 13,04853 |
Spezifikation Testergebnisse
Assay | Min. 99.0 % |
Solubility in alcohol | Passes test |
Free acidity | |
pH (20°C; 5 %) | 2.5 - 3.5 |
Heavy metals (as Pb) | Max. 5 ppm |
Ignition residue (SO4) | Max. 100 ppm |
Insolubility in water | Max. 50 ppm |
NH4 (Ammonium) | Max. 0.1 % |
SO4 (Sulphate) | Max. 20 ppm |
As (Arsenic) | Max. 1 ppm |
Cu (Copper) | Max. 10 ppm |
Fe (Iron) | |
Pb (Lead) | Max. 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...