meso-Tetraphenylporphin ≥99%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ200

247367-1G 247367-250MG
247367-1GEA 312 EUR
247367-1G 247367-250MG
meso-Tetraphenylporphin ≥99%, Sigma-Aldrich®
meso-Tetraphenylporphin
Formel: C₄₄H₃₀N₄
Dichte: 1,276 g/cm³ (20 °C)
Lagertemperatur: Room Temperature
MDL: MFCD00011680
CAS-Nummer: 917-23-7
EINECS: 213-025-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9db3ad3f15ef176ad5c2987e5e320fa439882d5&vid=ec7727426cb2cadc4ec56eb17800889046284e1b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Crystalline Powder
Infrared spectrum Conforms to Structure
Chlorin ≤0.1%
c = 0.03g/l in Toluene
Purity (HPLC) ≥99.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR