Glutaconaldehydedianil hydrochloride, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

247405-5G
247405-5GEA 251 EUR
247405-5G
Glutaconaldehydedianil hydrochloride, Sigma-Aldrich®
Glutaconaldehydedianil hydrochloride

Tricarbocyanine laser dye intermediate

Formel: C₁₇H₁₆N₂
Schmelzpunkt: 173…175 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012630
CAS-Nummer: 1497-49-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cb8b4946dffb2f1a22894272357d9b65929f026&vid=329dfd07b4b51f1ff2dc5882865a6e61f902f61c&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR