Xylol (Isomerengemisch) ≥98,5% (Xylene + Ethylbenzen Basis) ACS, Riedel-de Haën™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Dimethylbenzol
247642-20L
247642-1L-CB
247642-2.5L
247642-20LEA
735.69
EUR
247642-20L
247642-1L-CB
247642-2.5L
Xylol (Isomerengemisch) ≥98,5% (Xylene + Ethylbenzen Basis) ACS, Riedel-de Haën™
Xylol (Isomerengemisch)
ACS GENERAL USE SOLVENTS
- Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- Suitable for less demanding applications such as routine isocratic HPLC
- Variations and decrease issues traceable to solvent contamination
- All solvents are tested to meet ACS general use specifications
Formel:
C₆H₄(CH₃)₂ Molecular Weight: 106.17 g/mol Siedepunkt: 136…140 °C Schmelzpunkt: –34 °C Dichte: 0.87 g/cm³ Flammpunkt: 25 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077264 CAS-Nummer: 1330-20-7 UN: 1307 ADR: 3 III Merck Index: 13,10136 |
|
Spezifikation Testergebnisse
Assay Xylene isomers and Ethylbenzol (GC) | Min. 98.50 % |
Ethyl benzene (GC) | Max. 25 % |
Water (Karl Fischer) | Max. 0.05 % |
Non-volatile matter | Max. 0.002 % |
S-compounds (as S) | Max. 0.003 % |
APHA | Max. 10 |
Reaction against H₂SO₄ | complying |
Vapor Pressure | 18 mmHg ( 37.7 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...