Xylol (Isomerengemisch) ≥98,5% (Xylene + Ethylbenzen Basis) ACS, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

247642-20L 247642-1L-CB 247642-2.5L
247642-20LEA 735.69 EUR
247642-20L 247642-1L-CB 247642-2.5L
Xylol (Isomerengemisch) ≥98,5% (Xylene + Ethylbenzen Basis) ACS, Riedel-de Haën™
Xylol (Isomerengemisch)

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications

Formel: C₆H₄(CH₃)₂
Siedepunkt: 136…140 °C
Schmelzpunkt: –34 °C
Dichte: 0,87 g/cm³
Flammpunkt: 25 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00077264
CAS-Nummer: 1330-20-7
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay Xylene isomers and Ethylbenzol (GC) Min. 98.50 %
Ethyl benzene (GC) Max. 25 %
Water (Karl Fischer) Max. 0.05 %
Non-volatile matter Max. 0.002 %
S-compounds (as S) Max. 0.003 %
APHA Max. 10
Reaction against H₂SO₄ complying
Vapor Pressure 18 mmHg ( 37.7 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR