Xylylazo violet I, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 2-[2-Hydroxy-3-(2,4-xylylcarbamoyl)-1-naphthylazo]-1-phenol-4-sulphonic acid sodium salt, 1-Azo-2-hydroxy-3-(2,4-dimethylcarboxyanilido)naphthalene-1'-(2-hydroxybenzene-5-sulphonic acid) sodium salt, Xylidyl Blue I

M-Clarity™ Qualitätsstufe = MQ200

248266-1G
248266-1GEA 28.7 EUR
248266-1G
Xylylazo violet I, Sigma-Aldrich®
Xylylazo Violet I

Synonyms: 4-Hydroxy-3-[2-hydroxy-3-(2,4-dimethylphenylaminocarbonyl)-1-naphthylazobenzenesulfonic acid sodium salt; Magonsulfonate.

Formel: C₂₅H₂₁N₃O₆S
Molecular Weight: 513,50 g/mol
MDL: MFCD00003890
CAS-Nummer: 14936-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=974cad1e82c54f302879418eb6a70e483e4ca401&vid=60d0c01824f153d5335dc99a7bbe86ae50c16d7d&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (color) Red to purple, brown or black
Appearance (form) Crystal to powder
Infrared spectrum Conforms to structure
Wavelength (In ethanol, plus 0,08 M borate buffer) 252 - 262 nm
Extinction coefficient (typically) >13000
Wavelength 606 - 626 nm
Carbon 50,0 - 61,0%
Nitrogen 6,7 - 7,5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR