Chloride ionophore III, Selectophore™ Ionophor, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

24894-50MG
24894-50MGEA 513 EUR
24894-50MG
Chloride ionophore III, Selectophore™ Ionophor, Supelco®
Chloride ionophore III

Chloride selective ionophore for Cl--electrodes. ETH 9033 (chromoionophore III) may be used to monitor Cl- activity during the development of chloride sensitive probes encapsulated by biologically localized embedding.


It may also be used with Cl- ion sensitive ISFETs (ion-sensitive field-effect transistor) with photocured polyurethane-based polymer membrane to study sodium chloride solutions and to determine the chloride ions.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₃₂H₅₆Cl₂Hg₂O₂
MDL: MFCD00676703
CAS-Nummer: 178959-28-9
UN: 2025
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e5b159153509aa13516e32d4a5c7048b3e40179&vid=0a65df8c7b40964601803c3dec4f16bc0a886d5d&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR