N-Octadecanoyl nile blue, Selectophore™, Funktion getestet Ionophor, Supelco®

Lieferant: Merck
Gefahr

Synonyme: Chromoionophore I

M-Clarity™ Qualitätsstufe = MQ100

24901-25MG
24901-25MGEA 227 EUR
24901-25MG
N-Octadecanoyl nile blue, Selectophore™, Funktion getestet Ionophor, Supelco®
N-Octadecanoyl nile blue

Ionophore employed in solvent polymeric membrane electrodes for the assay of Cl- activity.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₃₈H₅₃N₃O₂
Molecular Weight: 583,85 g/mol
CAS-Nummer: 125829-24-5
UN: 2025
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42182e937e1435c5d7073b436318932779ab5124&vid=cf6ac9494e838afde19f7955a9ca0db14aef41b4&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR