Wasser, Cell Culture grade, tested USP Injection, sterile for sterile Water for Injection , Corning®

Lieferant: Corning

Synonyme: Wasserstoffoxid, Hydrogenoxid

25-055-LH 25-055-LG 25-055-LB 25-055-CVC 25-055-CM 25-055-CI 25-055-CV
MDTC25-055-LHEA 1350 EUR
MDTC25-055-LH MDTC25-055-LG MDTC25-055-LB MDTC25-055-CVC 25-055-CM MDTC25-055-CI 25-055-CV
Wasser, Cell Culture grade, tested USP Injection, sterile for sterile Water for Injection , Corning®
Wasser

Corning® Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications. Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications.


  • Complete Certificate of Analysis available for each production lot
  • Produced under the highest industry standards to ensure superior results
  • Meets WFI Standards


Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Mycoplasma Tested negative
Plating efficiency Pass
Oxidizable substances Solution remains faintly pink
Total organic carbon (before packaging) USP <643> < 500 ppb
Water conductivity (packaged water) USP <645> Meets USP Requirements
Water conductivity (before packaging) USP <645> Meets USP temperature/conductivity requirements
Endotoxin USP <85> ≤ 0.025 EU/mL
Sterility USP <71> Pass
Particulate matter USP <788> ≤ 25 particles/mL that are ≥ 10 μm in size
≤ 3 particles/mL that are ≥ 25 μm in size


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR