Wasser, erfüllt analytische Spezifikationen von Ph. Eur., USP zur Injizierung (WFI), Reagenz für die Zellkultur, steril, Corning®

Lieferant: Corning

25-065-LB 25-065-LG 25-065-LH
25-065-LBEA 493.16 EUR
25-065-LB 25-065-LG 25-065-LH
Wasser, erfüllt analytische Spezifikationen von Ph. Eur., USP zur Injizierung (WFI), Reagenz für die Zellkultur, steril, Corning®
Wasser

Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).


Technical Advantage: Wide range of applications in processes requiring very high levels of sterility USP/EP Certified Water is validated to the most recent specifications found in the United States Pharmacopeia (USP) and the European Pharmacopeia (EP). The specific testing is in accordance to testing for "Sterilized Water for Injection", (European Pharmacopeia), and "Sterile Water for Injection", (United States Pharmacopeia).

Formel: H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5

Bestellen

Spezifikation Testergebnisse

Mycoplasma Tested Negative
LM (plating effciency) Pass
Oxidizable Substances Pass
Total Organic Carbon (before packaging) USP <500 ppb
Water Conductivity (packaged water) USP Meets USP requirements
Water Conductivity (before packaging) USP Meets USP Temperature/Conductivity Requirements
Endotoxin USP ≤0.025 EU/ml
Sterility USP Pass
Acidity-Alkalinity Tested in Accordance with EP
Chlorides Pass
Nitrates ≤0.2 ppm
Sulphates Pass
Ammonium ≤0.2 ppm
Calcium and Magnesium Pass
Residue on Evaporation ≤3 mg (0.003)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR