Essigsäure (Eisessig) 99,7-100,5% USP, FCC, ACS, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

2502-14EA 50.2 EUR
2502-14 2502-27 2502-21 2502-22 2502-45 2502-46
Essigsäure (Eisessig) 99,7-100,5% USP, FCC, ACS, Macron Fine Chemicals™
Essigsäure

  • Food GMP manufactured product

Formel: H₃CCOOH
Siedepunkt: 118 °C (1013 hPa)
Schmelzpunkt: 17 °C
Dichte: 1,05 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00036152
CAS-Nummer: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product 99.7 - 100.5%
Meets ACS Reagent Chemical Requirements Passes Test
Meets F.C.C. Requirements ≥15.6 °C
Food GMP Manufactured Product ≤5 ppm
Meets U.S.P Requirements Passes Test
IMPORTANT: Material will freeze if stored below 17 °C (63°F). ≤0.005%
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Food Chemical Passes Test
Bulk Pharmaceutical Chemical 99.7 - 100.5%
USP - Assay (CH₃COOH) 99.7 - 100.5 %
USP - Chloride (Cl) (USP) Passes Test
USP - Congealing Temperature ≥15.6 °C
USP - Identification Passes Test
USP - Nonvolatile Residue ≤0.005 %
USP - Readily Oxidizable Substances Passes Test
USP - Sulfate (USP)(SO₄) Passes Test
FCC - Assay (CH₃COOH) 99.7 - 100.5 %
FCC - Identification Passes Test
FCC - Lead (Pb) ≤0.5 mg/kg
FCC - Nonvolatile Residue ≤0.005 %
FCC - Readily Oxidizable Substances Passes Test
FCC - Solidification Point ≥15.6 °C
ACS - Assay (CH₃COOH) (by GC) ≥99.7 %
ACS - Acetic Anhydride ((CH₃CO)₂O) ≤0.01 %
ACS - Chloride (Cl) ≤1 ppm
ACS - Color (APHA) ≤10
Dilution Test Passes Test
ACS - Heavy Metals (as Pb) (ACS) ≤0.5 ppm
Trace Impurities - ACS - Iron (Fe) ≤0.200 ppm
ACS - Residue after Evaporation ≤0.001 %
ACS - Substances Reducing Dichromate (ml) ≤0.40
ACS - Substances Reducing Permanganate Passes Test
ACS - Sulfate (SO₄) ≤1 ppm
ACS - Titrable Base (meq/g) ≤0.0004
Arsenic (As) ≤3 ppm
Copper (Cu) ≤0.00001 %
Freezing Point ≥16.0 °C
Nickel (Ni) ≤0.00001 %
Appearance Passes Test
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.
Residual Solvents: No Class 1, 2 or 3 residual solvents are used in or
produced by the production process.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR