Dihydrocytochalasin B, Calbiochem®

Lieferant: Merck Millipore Calbiochem

250225-1
250225-1EA 157 EUR
250225-1
Dihydrocytochalasin B, Calbiochem®
Dihydrocytochalasin B
Formel: C₂₉H₃₉NO₅
Molecular Weight: 481,63 g/mol
Schmelzpunkt: 198…203 °C
CAS-Nummer: 39156-67-7
EINECS: 254-324-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR