Cytochalasin B (aus Helminthosporium dematioideum), Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: 7(S),20(R)-Dihydroxy-16(R)-methyl-10-phenyl-24-oxa[14]cytochalasa-6(12),13(E),21(E)-triene-1,23-dione

250233-5
250233-5EA 286 EUR
250233-5
Cytochalasin B (aus Helminthosporium dematioideum), Millipore®
Cytochalasin B
Formel: C₂₉H₃₇NO₅
Molecular Weight: 479,62 g/mol
MDL: MFCD00077704
CAS-Nummer: 14930-96-2
EINECS: 239-000-2
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR