Cytochalasin D (aus Zygosporium mansoni), Calbiochem®
Lieferant: Merck Millipore Calbiochem
Synonyme:
7(S),18(R)-Dihydroxy-16(S),18-dimethyl-10-phenyl[11]cytochalasa-6(12),13(E),19(E)-trien-1,17-dion-21(R)-acetat
250255-1
250255-1EA
157
EUR
250255-1
Cytochalasin D (aus Zygosporium mansoni), Calbiochem®
Cytochalasin D
Formel:
C₃₀H₃₇NO₆ Molecular Weight: 507,63 g/mol |
MDL:
MFCD00077706 CAS-Nummer: 22144-77-0 EINECS: 244-804-1 UN: 1544 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...