Magnesiumsulfat, anhydrous 99, powder, BAKER ANALYZED®, J.T.Baker®

Lieferant: Avantor

Synonyme: MgSO4, Magnesium sulphate anhydrous

2506-01EA 178.11 EUR
2506-01 2506-05 2506-R
Magnesiumsulfat, anhydrous 99, powder, BAKER ANALYZED®, J.T.Baker®
Magnesium sulphate

The USP/NF Offical Compendia of Standards contains a section titled, "Reagents, Indicators, and Solutions" that lists the quality requirements of chemicals to be used in compendia testing. PharmaTest reagents are tested according to these requirements, relieving you of the need to do additional testing on your reagents. Products may be reagent or other grade, but are labeled as "Meets reagent specifications for testing USP/NF monographs." Test solutions are labeled as PharmaTest.

Formel: MgSO₄
Molecular Weight: 120.37 g/mol
Schmelzpunkt: 1127
Dichte: 2.65 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011110
CAS-Nummer: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use ≥99.0%
Assay (MgSO₄) (ignited basis) ≥99.0 %
Loss on Ignition ≤2.0 %
Chloride (Cl) ≤0.001 %
Nitrate (NO₃) ≤0.004 %
Ammonium (NH₄) ≤0.004 %
Calcium (Ca) ≤0.06 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Manganese (Mn) ≤0.001 %
Trace Impurities - Arsenic (As) ≤1 ppm
Meets Reagent Specifications for testing USP/NF monographs

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR