Daidzein, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-on, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyron, 4',7-Dihydroxyisoflavon, Daidzeol
251600-25
251600-25EA
70.6
EUR
251600-25
Daidzein, Millipore®
Daidzein
Formel:
C₁₅H₁₀O₄ Molecular Weight: 254.24 g/mol Siedepunkt: 513 Schmelzpunkt: 212…214 °C |
MDL:
MFCD00016954 CAS-Nummer: 486-66-8 EINECS: 207-635-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...