Magnesiumsulfat Hydrat 99,0-101,0%, GPR RECTAPUR®

Lieferant: VWR Chemicals

25169.295EA 66.3 EUR
25169.295
Magnesiumsulfat Hydrat 99,0-101,0%, GPR RECTAPUR®
Magnesiumsulfat Hydrat
Formel: MgSO₄·nH₂O
Dichte: 2,45 g/cm³ (20 °C)
MDL: MFCD00011110
CAS-Nummer: 22189-08-8

Bestellen

Spezifikation Testergebnisse

Assay (on anhydrous substance)
Heavy metals (as Pb) Max. 10 ppm
Cl (Chloride)
As (Arsenic) Max. 5 ppm
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR