5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 95% (Farbstoffgehalt)

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine

M-Clarity™ Qualitätsstufe = MQ100

252883-1G
252883-1GEA 119 EUR
252883-1G
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 95% (Farbstoffgehalt)
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin

5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine is an achiral porphyrin derivative. It forms chiral supramolecular assemblies via spontaneous symmetry breaking at the air or water interface.

Formel: C₄₈H₃₈N₄O₄
Molecular Weight: 734,86 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00012072
CAS-Nummer: 22112-78-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abdfbc28674f8565b7b68b5c02f46910b77bdc01&vid=abdfbc28674f8565b7b68b5c02f46910b77bdc01&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Purple
Appearance (Form), Powder, Crystals, Crystalline Powder and/or Chunks Conforms to Requirements
Infrared spectrum Conforms to Structure
Wavelength In Toluene 421 - 427 nm
Extinction Coefficient ≥350000
Carbon 73.7 - 83.2%
Nitrogen 7.2 - 8.2%
Purity (HPLC) ≥94.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR