D(-)-Mannitol zA

Lieferant: VWR Chemicals

Synonyme: (2R,3R,4R,5R)-(-)-1,2,3,4,5,6-Hexanhexol, D(-)-Mannit

25314.264EA 72.8 EUR
25314.264
D(-)-Mannitol zA
D(-)-Mannitol
Formel: HOH₂C(CH(OH))₄CH₂OH
Molecular Weight: 182.17 g/mol
Siedepunkt: 290…295 °C (4 hPa)
Schmelzpunkt: 164…169 °C
Dichte: 1.49 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064287
CAS-Nummer: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Reducing sugars Passes test
Acidity or alkalinity Passes test
Melting point 165 - 170 °C
Specific optical rotation (8 %; borate) 23 - 24 °
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 0.05 %
Loss on drying (100°C) Max. 0.3 %
Cl (Chloride) Max. 50 ppm
SO4 (Sulphate) Max. 100 ppm
As (Arsenic) Max. 2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR