δ-methrin 98 TLC_ASSAY_METHOD, solid_white, Calbiochem®

Lieferant: Merck Millipore Calbiochem
Gefahr

Synonyme: (S)-alpha-Cyano-3-phenoxybenzyl (1R,3R)-3-(2,2-Dibromovinyl)-2,2-dimethylcyclopropanecarboxylate, (1R,3R)-3-(2,2-Dibromovinyl)-2,2-dimethylcyclopropanecarboxylic acid (S)-alpha-Cyano-3-phenoxybenzyl ester, Deltamethrin

253300-10
253300-10EA 176 EUR
253300-10
δ-methrin 98 TLC_ASSAY_METHOD, solid_white, Calbiochem®
Deltamethrin

Deltamethrin is a synthetic type II pyrethroid that acts as a potent inhibitor of calcineurin (protein phosphatase 2B; IC50 = 100 pM). Also have varied effects on ion channels. Causes an increase in neurotransmitter release at synapses, and increases intrasynaptosomal Ca2+ levels.

Formel: C₂₂H₁₉Br₂NO₃
Molecular Weight: 505.206 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00870122
CAS-Nummer: 52918-63-5
UN: 2811
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR