Quecksilber(II)chlorid 99,5-100,5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS

Lieferant: VWR Chemicals
Gefahr

Synonyme: Quecksilberdichlorid, Sublimat

25384.298EA 984 EUR
25384.298
Quecksilber(II)chlorid 99,5-100,5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS
Quecksilber(II)chlorid
Formel: HgCl₂
Molecular Weight: 271,5 g/mol
Siedepunkt: 302 °C (1013 hPa)
Schmelzpunkt: 273 °C
Dichte: 5,44 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011041
CAS-Nummer: 7487-94-7
EINECS: 231-299-8
UN: 1624
ADR: 6.1,II
Merck Index: 13,05901

Bestellen

Spezifikation Testergebnisse

Assay (dried basis) 99.5 --> 100.5 %
Identification Passes test
Acidity or alkalinity Passes test
Appearance of solution Passes test
Mercury (I) chloride Passes test
Solution in ethyl ether Passes test
Loss on drying (under vacuum; 24 h) Max. 1.0 %
Residue after reduction Max. 0.02 %
Fe (Iron) Max. 20 ppm
Conforms to Reag. Ph.Eur. Passes test
Conforms to ACS Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR