Mangan(II)nitrat-Lösung 50 - 52%, BAKER ANALYZED®, J.T.Baker®

Lieferant: Avantor

2544-08EA 0 EUR
2544-08
Mangan(II)nitrat-Lösung 50 - 52%, BAKER ANALYZED®, J.T.Baker®
Mangan(II)nitrat
Formel: Mn(NO₃)₂
MDL: MFCD00011112
CAS-Nummer: 10377-66-9
UN: 3093
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Assay (Mn(NO₃)₂) 50.0 - 52.0%
Chloride (Cl) ≤0.002%
Sulphate (SO₄) ≤0.01%
Substances Not Precipitated by (NH₄)₂S (as SO₄) ≤0.2%
Other Heavy Metals (as Pb) ≤0.002%
Trace Impurities - Iron (Fe) ≤0.001%
Zinc (Zn) ≤0.02%
For Laboratory, Research or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR