Methyl-4-hydroxybenzoat 98,0-102,0% Ph. Eur.
Lieferant: VWR Chemicals
|
Achtung
|
Synonyme:
4-Hydroxybenzoesäuremethylester, Methylparaben
25604.290EA
149
EUR
25604.290
Methyl-4-hydroxybenzoat 98,0-102,0% Ph. Eur.
Methyl-4-hydroxybenzoat
Appearance: White or almost white, crystalline powder or colourless crystals.
Bitte beachten Not for use as Active Pharmaceutical Ingredient (API)
Formel:
HOC₆H₄CO₂CH₃ Molecular Weight: 152.15 g/mol Siedepunkt: 270…280 °C (1013 hPa) Schmelzpunkt: 125…128 °C Dichte: 1.1208 g/cm³ (137 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002352 CAS-Nummer: 99-76-3 EINECS: 202-785-7 Merck Index: 13,06129 |
|
Spezifikation Testergebnisse
Assay | 98,0 - 102,0 % |
Appearance | Conforms (see CoA/CoS) |
Melting point | 125 - 128 °C |
Identification (IR) | Passes test |
Appearance of solution | Passes test |
Acidity | Passes test |
4-Hydroxybenzoic acid | ≤ 0,5 % |
Unspecified impurities (for each) | ≤ 0,5 % |
Total impurities | ≤ 1,0 % |
Disregard limit | ≤ 0,1 % |
Sulphated ash | ≤ 0,1 % |
Residual solvents | Passes test |
Residual metal catalysts (skip lot) | |
As (Arsenic) | ≤ 2 ppm |
Cd (Cadmium) | ≤ 0.5 ppm |
Co (Cobalt) | ≤ 0.5 ppm |
Hg (Mercury) | ≤ 2 ppm |
Ni (Nickel) | ≤ 0.5 ppm |
Pb (Lead) | ≤ 2 ppm |
V (Vanadium) | ≤ 0.5 ppm |
Conforms to Ph.Eur. | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...