Ethylmethylketon ≥99,5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

25642.291EA 42 EUR
25642.291 25642.325 VWRC25642.460
Ethylmethylketon ≥99,5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens
Ethylmethylketon
Formel: C₂H₅COCH₃
Siedepunkt: 79,6 °C (1013 hPa)
Schmelzpunkt: –86 °C
Dichte: 0,8054 g/cm³ (20 °C)
Flammpunkt: –4 °C (DIN 51755 Part 1)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011648
CAS-Nummer: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3,II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay
IR Spectrum
Boiling point 79 - 80 °C
Colouration Max. 10 APHA
Density (20/4) 0.804 - 0.807
Density (20/20) 0.805 - 0.808
Acetone
Butan-2-ol
Methanol
2-Methylpropan-2-ol
Water
Al (Aluminium) Max. 0.2 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm
Conforms to ACS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR