Quecksilber(II)chlorid ≥99,5%, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Synonyme:
Quecksilberdichlorid, Sublimat
2594-01EA
0
EUR
2594-01
Quecksilber(II)chlorid ≥99,5%, BAKER ANALYZED® ACS, J.T.Baker®
Quecksilber(II)chlorid
Formel:
HgCl₂ Molecular Weight: 271.5 g/mol Siedepunkt: 302 Schmelzpunkt: 273 |
MDL:
MFCD00011041 CAS-Nummer: 7487-94-7 UN: 1624 ADR: 6.1 II Merck Index: 13,05901 |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use |
|
Assay (HgCl₂) | ≥99.5 % |
Residue after Reduction | ≤0.020 % |
Solution in Ether | Passes Test |
Iron (Fe) | ≤0.002 % |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...