Ethanol ≥96%, BAKER Ph. Eur., J.T.Baker®

Lieferant: Avantor

Synonyme: Ethyl alcohol

2607.91000EA 0 EUR
2607.91000 2607-38 2607-39
Ethanol ≥96%, BAKER Ph. Eur., J.T.Baker®
Ethanol
Formel: C₂H₅OH
Molecular Weight: 46.07 g/mol
Siedepunkt: 78.3
Schmelzpunkt: -117
Flammpunkt: 12
MDL: MFCD00003568
CAS-Nummer: 64-17-5
UN: 1170
ADR: 3 II
Merck Index: 12,03806

Bestellen

Spezifikation Testergebnisse

Meets Ph. Eur. Chemical Specifications.
Assay (C2H5OH) 92.6 - 95.2 % (m/m)
Acidity or Alkalinity (as CH3COOH) max. 30 ppm
Appearance passes test
Identification passes test
Relative Density 0.805 - 0.812
Residue on Evaporation max. 25 ppm (m/v)
Absorbance:
at 240 nm max. 0.40
at 250-260 nm max. 0.30
at 270-340 nm max. 0.10
Curve is smooth passes test
Volatile Impurities (by GC):
Acetaldehyde + Acetal max. 10 ppm (v/v)
Benzene max. 2 ppm (v/v)
Methanol (CH3OH) max. 200 ppm (v/v)
Total of other Impurities (based on surface area of internal standard 4-Methylpentan-2-ol) max. 300 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR