Salzsäure 10% in wässriger Lösung NF, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Hydrochloric acid solution, Muriatic acid

2608-08EA 0 EUR
2608-08 2608-22 2608-45 2608-02
Salzsäure 10% in wässriger Lösung NF, Macron Fine Chemicals™
Salzsäure
Formel: HCl
Molecular Weight: 36.46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 9.5 - 10.5
NF - Assay (HCl)(g/100ml) 9.5 - 10.5
NF - Identification (Cl) Passes Test
NF - Residue on Ignition ≤2 mg
NF - Free Bromine or Chlorine Passes Test
NF - Sulfate (SO₄) Passes Test
NF - Sulfite (SO₃) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR