Salzsäure 36,5 - 38,0% NF, FCC, ACS, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr


Veränderungsmanagement-Kategorie= R

2612-19EA 1789.12 EUR
2612-19 2612-06 2612-24 2612-44 2612-46 MACR2612-14
Salzsäure 36,5 - 38,0% NF, FCC, ACS, Macron Fine Chemicals™
Salzsäure
Formel: HCl
Siedepunkt: 110 °C (1013 hPa)
Schmelzpunkt: –30 °C
Dichte: 1,18 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets ACS Reagent Chemical Requirements
Meets F.C.C. Requirements
Meets N.F. Requirements 36.5 - 38.0 %
Food GMP Manufactured Product ≤ 3 ppm
Store below 25°C ≤ 0.010 ppm
CAUTION: For Manufacturing, processing or repackaging ≤ 0.005 %
Bulk Food Chemical ≤ 10
Bulk Pharmaceutical Chemical ≤ 5 ppm
ACS - Assay (HCl) (ACS) 36.5 - 38.0 %
ACS - Ammonium (NH₄) ≤ 3 ppm
ACS - Arsenic (As) ≤ 0.010 ppm
ACS - Bromide (Br) ≤ 0.005 %
ACS - Color (APHA) ≤ 10
ACS - Extractable Organic Substances ≤ 5 ppm
ACS - Free Chlorine (as Cl) ≤ 1 ppm
ACS - Residue after Ignition ≤ 5 ppm
ACS - Sulfate (SO₄) ≤ 1 ppm
ACS - Sulfite (SO₃) ≤ 1 ppm
ACS - Heavy Metals (as Pb) ≤ 1 ppm
Trace Impurities - ACS - Iron (Fe) ≤ 0.200 ppm
NF - Assay (HCl)(NF) 36.5 - 38.0 %
NF - Bromide or Iodide Passes Test
NF - Free Bromine or Chlorine Passes Test
NF - Identification Passes Test
NF - Residue on Ignition ≤ 0.008 %
NF - Sulfate (NF)(SO₄) Passes Test
NF - Sulfite (NF)(SO₃) Passes Test
FCC - Assay (HCl)(FCC) 36.5 - 38.0 %
FCC - Baume Degrees 22.7 - 23.4
FCC - Color (APHA) Passes Test
FCC - Identification Passes Test
FCC - Iron (Fe) ≤ 0.2 ppm
FCC - Lead (Pb) (mg/kg) ≤ 1
FCC - Mercury (Hg)(mg/kg) ≤ 0.10
FCC - Nonvolatile Residue ≤ 0.5 %
Total Organic Compounds - Non-Fluorine Containing (mg/kg) ≤ 5
Total Organic Compounds - Benzene (mg/kg) ≤ 0.05
Total Organic Compounds - Fluorinated Organic Compounds(mg/kg) ≤ 25
Total Organic Compounds - Oxidizing Substances ≤ 0.003 %
Reducing Substances ≤ 0.007 %
Specific Gravity 1.1856 - 1.1924
Sulfate (SO₄) ≤ 0.5 %
Appearance Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR