Natriumhydroxid, J.T.Baker®

Lieferant: Avantor

Synonyme: Caustic soda

2632-40EA 0 EUR
2632-40
Natriumhydroxid, J.T.Baker®
Sodium hydroxide
Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 1390
Schmelzpunkt: 323
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
UN: 1823
ADR: 8 II
Merck Index: 14,08627

Bestellen

Spezifikation Testergebnisse

Assay (NaOH) 16.0 - 18.0 %
Appearance (clear, colorless liquid) passes test
Identification (Na) passes test
Sodium Carbonate (Na2CO3) max. 0.6 %
pH at 25°C min. 11.0
Trace Impurities (in ppm)Chlorides (as Cl) max. 50
Trace Impurities (in ppm) Iron (Fe) max. 10
Trace Impurities (in ppm) Heavy Metals (as Pb) max. 20
Trace Impurities (in ppm) Sulfate (SO4) max. 50


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR