(2,5-Dimethoxyphenyl)acetyl chloride 99%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

264806-1G 264806-5G
264806-1GEA 100 EUR
264806-1G 264806-5G
(2,5-Dimethoxyphenyl)acetyl chloride 99%, Sigma-Aldrich®
(2,5-Dimethoxyphenyl)acetyl chloride
Formel: CH₁₁ClO₃
Siedepunkt: 118 °C/0,8 mmHg (Lit.)
Dichte: 1,227 g/ml (25 °C) (Lit.)
Flammpunkt: 108 °C (geschlossener Tiegel)
Lagertemperatur: Room Temperature
MDL: MFCD00012046
CAS-Nummer: 52711-92-9
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance (Colour); Yellow to Red to Brown Conforms to Requirements
Appearance (Form) Liquid
Infrared spectrum Conforms to Structure
Titration by AgNO3 After Oxygen Combustion: 98.5 - 101.5%
GC (area %) ≥98.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR