Dexamethason, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: 9-Fluor-11β,17α,21-trihydroxy-16α-methyl-pregna-1,4-dien-3,20-dion, Dexamethasonum

265005-100
265005-100EA 145 EUR
265005-100
Dexamethason, Millipore®
Dexamethason
Formel: C₂₂H₂₉FO₅
Molecular Weight: 392,47 g/mol
Schmelzpunkt: 268…271 °C
MDL: MFCD00064136
CAS-Nummer: 50-02-2
EINECS: 200-003-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR